CAS RN 2575516-45-7
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Famotidine Formaldehyde Adduct Maleate (2-[4-[2-(1,1-Dioxo-3,4-dihydro-2H-1,2,4,6-thiatriazin-5-yl)ethylsulfanylmethyl]-1,3-thiazol-2-yl]guanidine Maleate), Mikromol™
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.