CAS RN 2575516-52-6
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Phenylephrine Maleic Acid Adduct Hydrochloride (2-[6-(3-Hydroxyphenyl)-4-methyl-2-oxomorpholin-3-yl]acetic Acid Hydrochloride), Mikromol™
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.