CAS RN 2575516-76-4
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
(1R,4R)-1,2,3,4-Tetrahydro-4,6-dihydroxy-1-(3-hydroxyphenyl)-2-methylisoquinoline, Mikromol™
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.