CAS RN 257639-98-8
IUPAC Name:
N-butyl-N-ethyl-2,5-dimethyl-7-(2,4,6-trimethylphenyl)-7H-pyrrolo[2,3-d]pyrimidin-4-amine hydrochloride
Synonyms:
N-butyl-N-ethyl-2,5-dimethyl-7-(2,4,6-trimethylphenyl)pyrrolo[2,3-d]pyrimidin-4-amine hydrochloride
Molecular Weight (g/mol):
401
Molecular Formula:
C23H33ClN4
InChi Key:
ZWDLBNCJWNENFF-UHFFFAOYSA-N
SMILES:
Cl.CCCCN(CC)C1=C2C(C)=CN(C2=NC(C)=N1)C1=C(C)C=C(C)C=C1C