accessibility menu, dialog, popup

UserName

H2NOHNOHO

CAS RN 2577-40-4

IUPAC Name: 2-(2-amino-3-phenylpropanamido)-3-phenylpropanoic acid
Synonyms: phe-phe
Molecular Weight (g/mol): 312.37
Molecular Formula: C18H20N2O3
InChi Key: GKZIWHRNKRBEOH-UHFFFAOYNA-N
SMILES: NC(CC1=CC=CC=C1)C(=O)NC(CC1=CC=CC=C1)C(O)=O
Molecular Weight (g/mol) 
  • (3)
  • (2)
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (3)
  • (1)
  • (3)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)