CAS RN 2578-27-0
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Seleno-D,L-ethionine, TRC
CAS: 2578-27-0 Molecular Formula: C6 H13 N O2 Se Molecular Weight (g/mol): 210.13 IUPAC Name: 2-amino-4-ethylselanylbutanoic acid SMILES: CC[Se]CCC(N)C(=O)O