CAS RN 25781-92-4
IUPAC Name:
5-chloro-2-nitro-N-phenylaniline
Synonyms:
5-chloro-2-nitro-N-phenylaniline
Molecular Weight (g/mol):
248.67
Molecular Formula:
C12H9ClN2O2
InChi Key:
FPKHZBVGKMTUHB-UHFFFAOYSA-N
SMILES:
[O-][N+](=O)C1=CC=C(Cl)C=C1NC1=CC=CC=C1
1
–
3
of
3
results
1
5-Chloro-2-nitrodiphenylamine, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
5-Chloro-2-nitrodiphenylamine, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
5-Chloro-2-nitrodiphenylamine, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.