CAS RN 25784-91-2
IUPAC Name:
2-chloro-5-nitrobenzoyl chloride
Synonyms:
2-chloro-5-nitrobenzoyl chloride
Molecular Weight (g/mol):
220.01
Molecular Formula:
C7H3Cl2NO3
InChi Key:
OGLKKYALUKXVPQ-UHFFFAOYSA-N
SMILES:
[O-][N+](=O)C1=CC=C(Cl)C(=C1)C(Cl)=O
1
–
1
of
1
results
1
2-Chloro-5-nitrobenzoyl chloride, 96%, Thermo Scientific Chemicals
CAS: 25784-91-2 Molecular Formula: C7H3Cl2NO3 Molecular Weight (g/mol): 220.005 MDL Number: MFCD00059180 InChI Key: OGLKKYALUKXVPQ-UHFFFAOYSA-N Synonym: 2-chloro-5-nitrobenzoylchloride,benzoyl chloride, 2-chloro-5-nitro,acmc-1cq6z,2-chloro-5nitrobenzoyl chloride,2-chloro-5-nitro-benzoylchlorid,2-chloro-5-nitro-benzoic chloride,2-chloro-5-nitro-benzoyl chloride,benzoyl chloride,2-chloro-5-nitro,2-chloranyl-5-nitro-benzoyl chloride,2-chloro-5-nitrobenzoic acid chloride PubChem CID: 117637 IUPAC Name: 2-chloro-5-nitrobenzoyl chloride SMILES: C1=CC(=C(C=C1[N+](=O)[O-])C(=O)Cl)Cl