CAS RN 257877-44-4
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
3
of
3
results
1
8,8'-(1,4-Butanediyl)bis-8-azaspiro[4.5]decane-7,9-dione, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
8,8'-(Butane-1,4-diyl)bis(8-azaspiro[4.5]decane-7,9-dione), Mikromol™
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
3
8,8'-(1,4-Butanediyl)bis-8-azaspiro[4.5]decane-7,9-dione, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.