CAS RN 25830-77-7
IUPAC Name:
2-(2,6-dioxooxan-3-yl)-2,3-dihydro-1H-isoindole-1,3-dione
Synonyms:
2-(2,6-dioxooxan-3-yl)isoindole-1,3-dione
Molecular Weight (g/mol):
259.22
Molecular Formula:
C13H9NO5
InChi Key:
ICDLEMPZXFCQEB-UHFFFAOYNA-N
SMILES:
O=C1N(C2CCC(=O)OC2=O)C(=O)C2=CC=CC=C12
1
–
3
of
3
results
1
N-Phthaloyl-L-glutamic Anhydride, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
N-Phthaloyl-L-glutamic Anhydride, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
N-Phthaloyl-L-glutamic Anhydride, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.