accessibility menu, dialog, popup

UserName

ONOClNOCl

CAS RN 2587-01-1

IUPAC Name: 2,6-dichloro-4-nitropyridin-1-ium-1-olate
Synonyms: 2,6-dichloro-4-nitropyridin-1-ium-1-olate
Molecular Weight (g/mol): 208.98
Molecular Formula: C5H2Cl2N2O3
InChi Key: LWOLKOLHQCMQDE-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=CC(Cl)=[N+]([O-])C(Cl)=C1
Molecular Weight (g/mol) 
  • (1)
  • (3)
Quantity 
  • (1)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results