CAS RN 25895-62-9
IUPAC Name:
sodium cyanoboranuide
Synonyms:
sodium cyanoboranuide
Molecular Weight (g/mol):
62.84
Molecular Formula:
CH3BNNa
InChi Key:
CVDUGUOQTVTBJH-UHFFFAOYSA-N
SMILES:
[Na+].[BH3-]C#N
1
–
4
of
4
results
1
Sodium Cyanoborodeuteride, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
Sodium Cyanoborodeuteride, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
Sodium Cyanoborodeuteride, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
4
Sodium Cyanoborodeuteride, CDN
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.