CAS RN 25905-14-0
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
(±)-Lavandulyl Acetate, TRC
CAS: 25905-14-0 Molecular Formula: C12 H20 O2 Molecular Weight (g/mol): 196.29 Synonym: 4-Hexen-1-ol, 5-methyl-2-(1-methylethenyl)-, 1-acetate,4-Hexen-1-ol, 2-isopropenyl-5-methyl-, acetate (8CI),4-Hexen-1-ol, 5-methyl-2-(1-methylethenyl)-, acetate (9CI),(±)-Lavandulol acetate,(±)-Lavandulyl acetate,3-Acetoxymethyl-2,6-dimethyl-1,5-heptadiene,5-Methyl-2-(1-methylethenyl)-4-hexen-1-yl acetate,5-Methyl-2-(1-methylethenyl)-4-hexylene-1-ol acetate IUPAC Name: (5-methyl-2-prop-1-en-2-ylhex-4-enyl) acetate SMILES: CC(=CCC(COC(=O)C)C(=C)C)C