accessibility menu, dialog, popup

UserName

OHNHOBrBr

CAS RN 25952-74-3

IUPAC Name: 2,6-dibromo-4-[(hydroxyamino)methylidene]cyclohexa-2,5-dien-1-one
Synonyms: 2,6-dibromo-4-[(hydroxyamino)methylidene]cyclohexa-2,5-dien-1-one
Molecular Weight (g/mol): 294.93
Molecular Formula: C7H5Br2NO2
InChi Key: NWAZSJXQHSKZJO-UHFFFAOYSA-N
SMILES: ONC=C1C=C(Br)C(=O)C(Br)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results