CAS RN 259526-43-7
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
N-(2-Succinyl) Fluvoxamine, TRC
CAS: 259526-43-7 Molecular Formula: C19 H25 F3 N2 O6 Molecular Weight (g/mol): 434.41 Synonym: N-[2-[[(E)-[5-Methoxy-1-[4-(trifluoromethyl)phenyl]pentylidene]amino]oxy]ethyl]aspartic acid,(2RS)-2-[[2-[[[(1E)-5-methoxy-1-[4-(trifluoromethyl)phenyl]pentylidene]amino]oxy]ethyl]amino]butanedioic acid,Fluvoxamine Maleate Imp. C (EP) IUPAC Name: 2-[2-[(E)-[5-methoxy-1-[4-(trifluoromethyl)phenyl]pentylidene]amino]oxyethylamino]butanedioic acid SMILES: COCCCC\C(=N/OCCNC(CC(=O)O)C(=O)O)\c1ccc(cc1)C(F)(F)F