accessibility menu, dialog, popup

UserName

H3COOHONOO

CAS RN 2597-56-0

IUPAC Name: 2-methoxy-4-nitrobenzoic acid
Synonyms: 2-methoxy-4-nitrobenzoic acid
Molecular Weight (g/mol): 197.15
Molecular Formula: C8H7NO5
InChi Key: KPJXEWJRJKEOCD-UHFFFAOYSA-N
SMILES: COC1=CC(=CC=C1C(O)=O)[N+]([O-])=O
Molecular Weight (g/mol) 
  • (3)
  • (2)
Quantity 
  • (2)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results