CAS RN 259793-90-3
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
6-Chloro-3-hydroxypyrazine-2-carboxamide, TRC
CAS: 259793-90-3 Molecular Formula: C5H4N3O2Cl Molecular Weight (g/mol): 173.56 Synonym: Favipiravir Impurity 3 IUPAC Name: 5-chloro-2-oxo-1H-pyrazine-3-carboxamide SMILES: NC(=O)C1=NC(=CNC1=O)Cl