CAS RN 25991-27-9
IUPAC Name:
methyl 2-oxo-2H-pyran-3-carboxylate
Synonyms:
methyl 2-oxopyran-3-carboxylate
Molecular Weight (g/mol):
154.12
Molecular Formula:
C7H6O4
InChi Key:
GJVZWOMUTYNUCE-UHFFFAOYSA-N
SMILES:
COC(=O)C1=CC=COC1=O
1
–
1
of
1
results
1
Methyl 2-oxo-2H-pyran-3-carboxylate, 98%
CAS: 25991-27-9 Molecular Formula: C7H6O4 Molecular Weight (g/mol): 154.121 MDL Number: MFCD00075443 InChI Key: GJVZWOMUTYNUCE-UHFFFAOYSA-N Synonym: methyl 2-oxo-2h-pyran-3-carboxylate,3-carbomethoxy-2-pyrone,methyl 2-pyrone-3-carboxylate,2-oxo-2h-pyran-3-carboxylic acid methyl ester,2h-pyran-3-carboxylic acid, 2-oxo-, methyl ester,methoxycarbonyl pyranone,acmc-1clg5,ksc496k6b PubChem CID: 117704 IUPAC Name: methyl 2-oxopyran-3-carboxylate SMILES: COC(=O)C1=CC=COC1=O