CAS RN 2602-34-8
IUPAC Name:
triethoxy({3-[(oxiran-2-yl)methoxy]propyl})silane
Synonyms:
triethoxy[3-(oxiran-2-ylmethoxy)propyl]silane
Molecular Weight (g/mol):
278.42
Molecular Formula:
C12H26O5Si
InChi Key:
JXUKBNICSRJFAP-UHFFFAOYNA-N
SMILES:
CCO[Si](CCCOCC1CO1)(OCC)OCC
1
–
1
of
1
results
1
(3-Glycidoxypropyl)triethoxysilane, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.