CAS RN 26021-57-8
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
3,4-Dihydro-2H-benzo[b][1,4]oxazin-6-ol, TRC
CAS: 26021-57-8 Molecular Formula: C8 H9 N O2 Molecular Weight (g/mol): 151.16 IUPAC Name: 3,4-dihydro-2H-1,4-benzoxazin-6-ol SMILES: Oc1ccc2OCCNc2c1