CAS RN 2602451-77-2
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Methyl 3-[[(2RS)-2-(Dipropylamino)propanoyl]amino]-4-methylthiophene-2-carboxylate Hydrochloride (Dipropylarticaine Hydrochloride), Mikromol™
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.