CAS RN 260779-88-2
IUPAC Name:
4-amino-5-chloro-N-{1-[3-(4-fluorophenoxy)propyl]-3-methoxypiperidin-4-yl}-2-methoxybenzamide hydrate
Synonyms:
cisapride hydrate
Molecular Weight (g/mol):
483.97
Molecular Formula:
C23H31ClFN3O5
InChi Key:
QBYYXIDJOFZORM-UHFFFAOYNA-N
SMILES:
O.COC1CN(CCCOC2=CC=C(F)C=C2)CCC1NC(=O)C1=CC(Cl)=C(N)C=C1OC