accessibility menu, dialog, popup

UserName

H3CHNOOCH3H3C

CAS RN 26078-25-1

IUPAC Name: 7-(ethylamino)-4,6-dimethyl-2H-chromen-2-one
Synonyms: 7-(ethylamino)-4,6-dimethylchromen-2-one
Molecular Weight (g/mol): 217.27
Molecular Formula: C13H15NO2
InChi Key: QZXAEJGHNXJTSE-UHFFFAOYSA-N
SMILES: CCNC1=C(C)C=C2C(C)=CC(=O)OC2=C1
Color 
  • (1)
Grade 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)