accessibility menu, dialog, popup

UserName

OOOHOOOHOHOHOHOHOH

CAS RN 260804-65-7

IUPAC Name: 2-{[6-(2-cyclohexylethoxy)-4,5-dihydroxy-2-(hydroxymethyl)oxan-3-yl]oxy}-6-(hydroxymethyl)oxane-3,4,5-triol
Synonyms: 2-{[6-(2-cyclohexylethoxy)-4,5-dihydroxy-2-(hydroxymethyl)oxan-3-yl]oxy}-6-(hydroxymethyl)oxane-3,4,5-triol
Molecular Weight (g/mol): 452.5
Molecular Formula: C20H36O11
InChi Key: RHXPDNGQJSXOMW-UHFFFAOYNA-N
SMILES: OCC1OC(OC2C(O)C(O)C(OCCC3CCCCC3)OC2CO)C(O)C(O)C1O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results