CAS RN 2609-23-6
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
2,6-Dimethyl-trans-2,6-octadiene, TRC
CAS: 2609-23-6 Molecular Formula: C10H18 Molecular Weight (g/mol): 138.25 Synonym: (6E)-2,6-Dimethyl-2,6-octadiene,(E)-2,6-Dimethyl-2,6-octadiene,Deoxygeraniol,trans-2,6-Dimethyl-2,6-octadiene SMILES: C/C(CC/C=C(C)/C)=C\C