CAS RN 26103-97-9
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Phlorisovalerophenone, TRC
CAS: 26103-97-9 Molecular Formula: C11 H14 O4 Molecular Weight (g/mol): 210.23 Synonym: 1-Butanone, 3-methyl-1-(2,4,6-trihydroxyphenyl)-,Butyrophenone, 2',4',6'-trihydroxy-3-methyl- (8CI),Phlorobutyrophenone, 3-methyl- (6CI),3-Methyl-1-(2,4,6-trihydroxyphenyl)-1-butanone,2,4,6-Trihydroxyisovalerophenone,2',4',6'-Trihydroxyisovalerophenone,Isovaleroylphloroglucinol,Isovalerylphloroglucinol,Phlorisovalerophenone,Phloroisovalerophenone,Preleptospermone IUPAC Name: 3-methyl-1-(2,4,6-trihydroxyphenyl)butan-1-one SMILES: CC(C)CC(=O)c1c(O)cc(O)cc1O