CAS RN 261356-49-4
IUPAC Name:
potassium tetracyanoboranuide
Synonyms:
potassium tetracyanoboranuide
Molecular Weight (g/mol):
153.98
Molecular Formula:
C4BKN4
InChi Key:
SQFZXQOCCKUPEC-UHFFFAOYSA-N
SMILES:
[K+].N#C[B-](C#N)(C#N)C#N
1
–
1
of
1
results
1
Potassium Tetracyanoborate, TRC
CAS: 261356-49-4 Molecular Formula: C4 B N4 . K Molecular Weight (g/mol): 153.979 IUPAC Name: potassium;tetracyanoboranuide SMILES: [K+].N#C[B-](C#N)(C#N)C#N