accessibility menu, dialog, popup

UserName

CH3NCH3NCH3CH3O(E)(E)

CAS RN 261360-66-1

IUPAC Name: (2E,5E)-2,5-bis({[4-(diethylamino)phenyl]methylidene})cyclopentan-1-one
Synonyms: (2E,5E)-2,5-bis({[4-(diethylamino)phenyl]methylidene})cyclopentan-1-one
Molecular Weight (g/mol): 402.58
Molecular Formula: C27H34N2O
InChi Key: KKLBPVXKMBLCQX-BLVCXSLXSA-N
SMILES: CCN(CC)C1=CC=C(\C=C2/CC\C(=C/C3=CC=C(C=C3)N(CC)CC)C2=O)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results