CAS RN 2614335-77-0
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
p-SCN-Bn-DTPA Trihydrochloride Salt (>90%), TRC
CAS: 2614335-77-0 Molecular Formula: C22H28N4O10S • 3(HCl) Molecular Weight (g/mol): 649.92 Synonym: N-[2-[Bis(carboxymethyl)amino]ethyl]-N-[(2S)-2-[bis(carboxymethyl)amino]-3-(4-isothiocyanatophenyl)propyl]glycine Trihydrochloride,(S)-1-(4-Isothiocyanatobenzyl)diethylenetriamine-N,N,N',N' ',N' '-pentaacetic Acid Trihydrochloride; IUPAC Name: (S)-2,2'-((2-((2-(bis(carboxymethyl)amino)-3-(4-isothiocyanatophenyl)propyl)(carboxymethyl)amino)ethyl)azanediyl)diacetic acid hydrochloride SMILES: OC(CN([C@@H](CC1=CC=C(C=C1)N=C=S)CN(CCN(CC(O)=O)CC(O)=O)CC(O)=O)CC(O)=O)=O.Cl