CAS RN 261733-18-0
IUPAC Name:
4,6-bis(diphenylphosphanyl)-10H-phenoxazine
Synonyms:
4,6-bis(diphenylphosphanyl)-10H-phenoxazine
Molecular Weight (g/mol):
551.57
Molecular Formula:
C36H27NOP2
InChi Key:
HSWZLYXRAOXOLL-UHFFFAOYSA-N
SMILES:
N1C2=C(OC3=C1C=CC=C3P(C1=CC=CC=C1)C1=CC=CC=C1)C(=CC=C2)P(C1=CC=CC=C1)C1=CC=CC=C1
1
–
1
of
1
results
1
4,6-Bis(diphenylphosphino)phenoxazine, 98+%
CAS: 261733-18-0 Molecular Formula: C36H27NOP2 Molecular Weight (g/mol): 551.57 MDL Number: MFCD03788937 InChI Key: HSWZLYXRAOXOLL-UHFFFAOYSA-N Synonym: 4,6-bis diphenylphosphino phenoxazine,4,6-bis diphenylphosphino-10h-phenoxazine,nixantphos,10h-phenoxazine, 4,6-bis diphenylphosphino,4,6-bis diphenylphosphanyl-10h-phenoxazine,nixanthphos,n-xantphos,acmc-209gp1,bis diphenylphosphino phenoxazine,4,6-bis diphenylphosphino-phenoxazine PubChem CID: 3255346 IUPAC Name: (6-diphenylphosphanyl-10H-phenoxazin-4-yl)-diphenylphosphane SMILES: N1C2=C(OC3=C1C=CC=C3P(C1=CC=CC=C1)C1=CC=CC=C1)C(=CC=C2)P(C1=CC=CC=C1)C1=CC=CC=C1