accessibility menu, dialog, popup

UserName

H3COFBrCl

CAS RN 261762-34-9

IUPAC Name: 5-bromo-1-chloro-3-fluoro-2-methoxybenzene
Synonyms: 5-bromo-1-chloro-3-fluoro-2-methoxybenzene
Molecular Weight (g/mol): 239.47
Molecular Formula: C7H5BrClFO
InChi Key: IKWHDHGNXXVBHZ-UHFFFAOYSA-N
SMILES: COC1=C(Cl)C=C(Br)C=C1F
Molecular Weight (g/mol) 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results