CAS RN 2618-96-4
IUPAC Name:
N-(benzenesulfonyl)benzenesulfonamide
Synonyms:
N-(benzenesulfonyl)benzenesulfonamide
Molecular Weight (g/mol):
297.34
Molecular Formula:
C12H11NO4S2
InChi Key:
OVQABVAKPIYHIG-UHFFFAOYSA-N
SMILES:
O=S(=O)(NS(=O)(=O)C1=CC=CC=C1)C1=CC=CC=C1