accessibility menu, dialog, popup

UserName

CH3BrOHOHO

CAS RN 261904-39-6

IUPAC Name: 3-bromo-2-hydroxy-2-methylpropanoic acid
Synonyms: 3-bromo-2-hydroxy-2-methylpropanoic acid
Molecular Weight (g/mol): 183
Molecular Formula: C4H7BrO3
InChi Key: HBJAYXGUOOININ-UHFFFAOYNA-N
SMILES: CC(O)(CBr)C(O)=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results