accessibility menu, dialog, popup

UserName

CH3OHOCH3OHOPPRu(?)

CAS RN 261948-85-0

IUPAC Name: [2'-(diphenylphosphanyl)-[1,1'-binaphthalen]-2-yl]diphenylphosphane bis(acetic acid) ruthenium
Synonyms: bis(acetic acid) binap ruthenium
Molecular Weight (g/mol): 843.86
Molecular Formula: C48H40O4P2Ru
InChi Key: XANQJWCSQJDBEA-UHFFFAOYSA-N
SMILES: [Ru].CC(O)=O.CC(O)=O.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=C2C=CC=CC2=C1C1=C2C=CC=CC2=CC=C1P(C1=CC=CC=C1)C1=CC=CC=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (1)
  • (1)