CAS RN 261952-05-0
IUPAC Name:
4-methyl-2-(trifluoromethyl)benzonitrile
Synonyms:
4-methyl-2-(trifluoromethyl)benzonitrile
Molecular Weight (g/mol):
185.15
Molecular Formula:
C9H6F3N
InChi Key:
WCZWEODOIUKVHX-UHFFFAOYSA-N
SMILES:
CC1=CC(=C(C=C1)C#N)C(F)(F)F
1
–
1
of
1
results
1
4-Methyl-2-(trifluoromethyl)benzonitrile, 98%
CAS: 261952-05-0 Molecular Formula: C9H6F3N Molecular Weight (g/mol): 185.15 MDL Number: MFCD01631596 InChI Key: WCZWEODOIUKVHX-UHFFFAOYSA-N Synonym: 4-methyl-2-trifluoromethyl benzonitrile,4-methyl-2-trifluoromethyl benzenecarbonitrile,2-cyano-5-methylbenzotrifluoride,2-trifluoromethyl-p-tolunitrile,4-cyano-3-trifluoromethyl toluene,benzonitrile,4-methyl-2-trifluoromethyl PubChem CID: 2775597 IUPAC Name: 4-methyl-2-(trifluoromethyl)benzonitrile SMILES: CC1=CC(=C(C=C1)C#N)C(F)(F)F