CAS RN 261952-19-6
IUPAC Name:
4-(bromomethyl)-1-methyl-2-(trifluoromethyl)benzene
Synonyms:
4-(bromomethyl)-1-methyl-2-(trifluoromethyl)benzene
Molecular Weight (g/mol):
253.06
Molecular Formula:
C9H8BrF3
InChi Key:
DEGVSFJBDJUTBP-UHFFFAOYSA-N
SMILES:
CC1=C(C=C(CBr)C=C1)C(F)(F)F
1
–
1
of
1
results
1
4-Methyl-3-(trifluoromethyl)benzyl bromide, 97%, Thermo Scientific™
CAS: 261952-19-6 Molecular Formula: C9H8BrF3 Molecular Weight (g/mol): 253.06 MDL Number: MFCD01631612 InChI Key: DEGVSFJBDJUTBP-UHFFFAOYSA-N Synonym: 4-bromomethyl-1-methyl-2-trifluoromethyl benzene,4-methyl-3-trifluoromethyl benzyl bromide,acmc-1ciaw,4-methyl-3-trifluoromethyl benzylbromide,benzene,4-bromomethyl-1-methyl-2-trifluoromethyl PubChem CID: 2775627 IUPAC Name: 4-(bromomethyl)-1-methyl-2-(trifluoromethyl)benzene SMILES: CC1=C(C=C(CBr)C=C1)C(F)(F)F