CAS RN 26197-93-3
IUPAC Name:
4-bromobenzene-1-carbothioamide
Synonyms:
4-bromobenzenecarbothioamide
Molecular Weight (g/mol):
216.1
Molecular Formula:
C7H6BrNS
InChi Key:
AIPANIYQEBQYGC-UHFFFAOYSA-N
SMILES:
NC(=S)C1=CC=C(Br)C=C1
1
–
1
of
1
results
1
4-Bromothiobenzamide, 97%
CAS: 26197-93-3 Molecular Formula: C7H6BrNS Molecular Weight (g/mol): 216.096 MDL Number: MFCD04114360 InChI Key: AIPANIYQEBQYGC-UHFFFAOYSA-N Synonym: 4-bromothiobenzamide,4-bromo-thiobenzamide,p-bromothiobenzamide,4-bromobenzothioamide,benzenecarbothioamide, 4-bromo,4-bromobenzene-1-carbothioamide,amino 4-bromophenyl methane-1-thione,acmc-1cehr,4-bromanylbenzenecarbothioamide PubChem CID: 1549289 IUPAC Name: 4-bromobenzenecarbothioamide SMILES: C1=CC(=CC=C1C(=S)N)Br