CAS RN 26225-79-6
IUPAC Name:
2-ethoxy-3,3-dimethyl-2,3-dihydro-1-benzofuran-5-yl methanesulfonate
Synonyms:
ethofumesate
Molecular Weight (g/mol):
286.34
Molecular Formula:
C13H18O5S
InChi Key:
IRCMYGHHKLLGHV-UHFFFAOYNA-N
SMILES:
CCOC1OC2=CC=C(OS(C)(=O)=O)C=C2C1(C)C
1
–
5
of
5
results
2
Ethofumesate, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.