accessibility menu, dialog, popup

UserName

H3COON(R)CH3(S)NFFFFFFOOCH3FFF

CAS RN 262352-17-0

IUPAC Name: ethyl (2R,4S)-4-({[3,5-bis(trifluoromethyl)phenyl]methyl}(methoxycarbonyl)amino)-2-ethyl-6-(trifluoromethyl)-1,2,3,4-tetrahydroquinoline-1-carboxylate
Synonyms: torcetrapib
Molecular Weight (g/mol): 600.48
Molecular Formula: C26H25F9N2O4
InChi Key: CMSGWTNRGKRWGS-NQIIRXRSSA-N
SMILES: CCOC(=O)N1[C@H](CC)C[C@H](N(CC2=CC(=CC(=C2)C(F)(F)F)C(F)(F)F)C(=O)OC)C2=CC(=CC=C12)C(F)(F)F
Quantity 
  • (2)
  • (1)
  • (1)
Color 
  • (3)
Grade 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results