CAS RN 262368-30-9
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
N-(4-Aminophenyl)-N-methyl-2-(4-methylpiperazin-1-yl)acetamide, TRC
CAS: 262368-30-9 Molecular Formula: C14H22N4O Molecular Weight (g/mol): 262.35 Synonym: N-(4-Aminophenyl)-N,4-dimethyl-1-piperazineacetamide,N-[[(4-Methylpiperazin-1-yl)methyl]carbonyl]-N-methyl-p-phenylenediamine IUPAC Name: N-(4-aminophenyl)-N-methyl-2-(4-methylpiperazin-1-yl)acetamide SMILES: CN1CCN(CC1)CC(=O)N(C)C2=CC=C(C=C2)N