CAS RN 2629-54-1
IUPAC Name:
(2S)-2-azaniumyl-3-(3-fluorophenyl)propanoate
Synonyms:
(2S)-2-ammonio-3-(3-fluorophenyl)propanoate
Molecular Weight (g/mol):
183.18
Molecular Formula:
C9H10FNO2
InChi Key:
VWHRYODZTDMVSS-QMMMGPOBSA-N
SMILES:
[NH3+][C@@H](CC1=CC=CC(F)=C1)C([O-])=O
1
–
1
of
1
results
1
3-Fluoro-DL-phenylalanine, 98%
CAS: 2629-54-1 Molecular Formula: C9H10FNO2 Molecular Weight (g/mol): 183.18 MDL Number: MFCD00004273,MFCD00066450 InChI Key: VWHRYODZTDMVSS-QMMMGPOBSA-N Synonym: 3-fluoro-dl-phenylalanine,3-fluorophenylalanine,2-amino-3-3-fluorophenyl propanoic acid,m-fluorophenylalanine,dl-3-fluorophenylalanine,h-dl-phe 3-f-oh,m-fluoro-dl-phenylalanine,alanine, 3-fluorophenyl,3-fluoro-3-phenylalanine,dl-phenylalanine, 3-fluoro PubChem CID: 9976 IUPAC Name: 2-amino-3-(3-fluorophenyl)propanoic acid SMILES: [NH3+][C@@H](CC1=CC=CC(F)=C1)C([O-])=O