accessibility menu, dialog, popup

UserName

PPPClCo

CAS RN 26305-75-9

IUPAC Name: tris(triphenylphosphane) cobalt chloride
Synonyms: tris(triphenylphosphine) cobalt chloride
Molecular Weight (g/mol): 881.26
Molecular Formula: C54H45ClCoP3
InChi Key: LNEYYSPXDKDADL-UHFFFAOYSA-M
SMILES: [Cl-].[Co].C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results