accessibility menu, dialog, popup

UserName

NOHH2NONFFF(E/Z)

CAS RN 263161-34-8

IUPAC Name: N'-hydroxy-4-{[5-(trifluoromethyl)pyridin-2-yl]oxy}benzene-1-carboximidamide
Synonyms: N'-hydroxy-4-{[5-(trifluoromethyl)pyridin-2-yl]oxy}benzenecarboximidamide
Molecular Weight (g/mol): 297.24
Molecular Formula: C13H10F3N3O2
InChi Key: RHAVFJMKHOLIST-UHFFFAOYSA-N
SMILES: NC(=NO)C1=CC=C(OC2=NC=C(C=C2)C(F)(F)F)C=C1
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results