accessibility menu, dialog, popup

UserName

CAS RN 263349-73-1

SOONH2FBr

CAS RN 263349-73-1

IUPAC Name: 4-bromo-3-fluorobenzene-1-sulfonamide
Synonyms: 4-bromo-3-fluorobenzenesulfonamide
Molecular Weight (g/mol): 254.07
Molecular Formula: C6H5BrFNO2S
InChi Key: QTICZSSQSQDJEX-UHFFFAOYSA-N
SMILES: NS(=O)(=O)C1=CC(F)=C(Br)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results