CAS RN 2636-52-4
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
L-Primapterin, TRC
CAS: 2636-52-4 Molecular Formula: C9H11N5O3 Molecular Weight (g/mol): 237.22 Synonym: 2-Amino-7-[(1R,2S)-1,2-dihydroxypropyl]-4(3H)-pteridinone,L-erythro- 1-(2-Amino-4-hydroxy-7-pteridinyl)-1,2-propanediol,2-Amino-7-[(1R,2S)- 1,2-dihydroxypropyl]-4(1H)-pteridinone,7-Biopterin,Primapterin; SMILES: C[C@@H]([C@@H](C1=CN=C2C(=N1)NC(=NC2=O)N)O)O