accessibility menu, dialog, popup

UserName

CAS RN 2637-34-5

SNH

CAS RN 2637-34-5

IUPAC Name: 1,2-dihydropyridine-2-thione
Synonyms: 2(1H)-pyridinethione
Molecular Weight (g/mol): 111.16
Molecular Formula: C5H5NS
InChi Key: WHMDPDGBKYUEMW-UHFFFAOYSA-N
SMILES: S=C1NC=CC=C1
Molecular Weight (g/mol) 
  • (4)
  • (3)
Quantity 
  • (2)
  • (1)
  • (2)
  • (2)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (1)
  • (4)
  • (3)
  • (2)
  • (1)
  • (2)
  • (2)
  • (3)