CAS RN 26447-85-8
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
2
of
2
results
1
Methyl Di(2-thienylglycolate), TRC
CAS: 26447-85-8 Molecular Formula: C11 H10 O3 S2 Molecular Weight (g/mol): 254.33 Synonym: Methyl 2-Hydroxy-2,2-dithiophen-2-ylacetate,Tiotropium Bromide Monohydrate Imp. E (EP) IUPAC Name: methyl 2-hydroxy-2,2-dithiophen-2-ylacetate SMILES: COC(=O)C(O)(c1cccs1)c2cccs2
2
Tiotropium EP Impurity E, Mikromol™
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.