accessibility menu, dialog, popup

UserName

CH3NH3CONHNNOClClHCl

CAS RN 264618-38-4

IUPAC Name: 1-(1-{3-[1-benzoyl-3-(3,4-dichlorophenyl)piperidin-3-yl]propyl}-4-phenylpiperidin-4-yl)-3,3-dimethylurea hydrochloride
Synonyms: 1-(1-{3-[1-benzoyl-3-(3,4-dichlorophenyl)piperidin-3-yl]propyl}-4-phenylpiperidin-4-yl)-3,3-dimethylurea hydrochloride
Molecular Weight (g/mol): 658.11
Molecular Formula: C35H43Cl3N4O2
InChi Key: HYPKKQPFHSNZBY-UHFFFAOYNA-N
SMILES: Cl.CN(C)C(=O)NC1(CCN(CCCC2(CCCN(C2)C(=O)C2=CC=CC=C2)C2=CC(Cl)=C(Cl)C=C2)CC1)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results