CAS RN 264624-38-6
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
27-Deoxyactein, TRC
CAS: 264624-38-6 Molecular Formula: C37 H56 O10 Molecular Weight (g/mol): 660.83 Synonym: β-D-Xylopyranoside, (3β,12β,16β,23S,24R,25R)-12-(acetyloxy)-16,23:23,26:24,25-triepoxy-9,19-cyclolanostan-3-yl (9CI, ACI),(3β,12β,16β,23S,24R,25R)-12-(Acetyloxy)-16,23:23,26:24,25-triepoxy-9,19-cyclolanostan-3-yl β-D-xylopyranoside (ACI),26-Deoxyactein,27-Deoxyactein SMILES: C[C@@H]1C[C@]2(OC[C@@]3(C)O[C@@H]23)O[C@H]4C[C@@]5(C)[C@@H]6CC[C@H]7C(C)(C)[C@H](CC[C@@]78C[C@@]68C[C@@H](OC(=O)C)[C@]5(C)[C@@H]14)O[C@@H]9OC[C@@H](O)[C@H](O)[C@H]9O