accessibility menu, dialog, popup

UserName

CH3H3C

CAS RN 26472-00-4

IUPAC Name: 4,9-dimethyltricyclo[5.2.1.0²,⁶]deca-3,8-diene
Synonyms: 4,9-dimethyltricyclo[5.2.1.0²,⁶]deca-3,8-diene
Molecular Weight (g/mol): 160.26
Molecular Formula: C12H16
InChi Key: IYQYZZHQSZMZIG-UHFFFAOYNA-N
SMILES: CC1=CC2C(C1)C1CC2C(C)=C1
Boiling Point 
  • (3)
  • (3)
Percent Purity 
  • (3)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Grade 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (3)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (3)